missing translation for 'onlineSavingsMsg'
Learn More

Apremilast, MedChemExpress

Product Code. 16494978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494978 10 mg 10mg
16493276 25 mg 25mg
16485746 100 mg 100mg
16461227 50 mg 50mg
16417717 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494978 Supplier Medchem Express Supplier No. HY12085
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Apremilast (CC-10004) is an orally available inhibitor of type-4 cyclic nucleotide phosphodiesterase (PDE-4) with an IC50 of 74 nM. Apremilast inhibits TNF-α release by lipopolysaccharide (LPS) with an IC50 of 104 nM.

Apremilast (CC-10004) is an orally available inhibitor of type-4 cyclic nucleotide phosphodiesterase (PDE-4) with an IC50 of 74 nM. Apremilast inhibits TNF-α release by lipopolysaccharide (LPS) with an IC50 of 104 nM.

Specifications

Chemical Name or Material Apremilast
CAS 608141-41-9
Color White
Purity Grade Notes Research
Molecular Formula C22H24N2O7S
Quantity 10 mg
Synonym CC-10004
Solubility Information DMSO : 44 mg/mL (95.55 mM; Need ultrasonic)
SMILES CC(NC1=CC=CC(C(N2[C@@H](C3=CC=C(OC)C(OCC)=C3)CS(=O)(C)=O)=O)=C1C2=O)=O
Molecular Weight (g/mol) 460.5
Formula Weight 460.5
Percent Purity 98.99%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.