missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aprataxin Recombinant Rabbit Monoclonal Antibody (24GB6830)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen Aprataxin
Clone 24GB6830
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275338
GREENER_CHOICE
View Documents
Invitrogen™
MA556965
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274837
View Documents
Invitrogen™
MA556966
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aprataxin Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the histidine triad (HIT) superfamily, some of which have nucleotide-binding and diadenosine polyphosphate hydrolase activities. The encoded protein may play a role in single-stranded DNA repair. Mutations in this gene have been associated with ataxia-ocular apraxia. Multiple transcript variants encoding distinct isoforms have been identified for this gene, however, the full length nature of some variants has not been determined.
Specifications

Specifications

Aprataxin
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
APTX
2410016G21Rik; AA388047; AOA; AOA1; Aprataxin; aprataxin isoform a; Aptx; APTX protein; AXA1; EAOH; EOAHA; FHA-HIT; Forkhead-associated domain histidine triad-like protein; forkhead-associated domain histidine-triad like protein
APTX
Primary
Affinity chromatography
24GB6830
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q7Z2E3
54840
A synthesized peptide derived from human ApRataxin (aa 147-160).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.