missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aprataxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-534
This item is not returnable.
View return policy
Description
Aprataxin Polyclonal specifically detects Aprataxin in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Aprataxin | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| AOA1EAOH, AOAMGC1072, aprataxin, ataxia 1, early onset with hypoalbuminemia, AXA1, EC 3.-, EOAHA, FHA-HIT, FLJ20157, Forkhead-associated domain histidine triad-like protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54840 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot, Immunoprecipitation 1 - 4 ug/mg lysate | |
| Q7Z2E3 | |
| APTX | |
| The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Aprataxin using the numbering given in SwissProt Entry Q7Z2E3 (GeneID 54840). | |
| 100 μL | |
| Primary | |
| NB100-534 is specific for human Aprataxin protein. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction