missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aprataxin Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-534UV
This item is not returnable.
View return policy
Description
Aprataxin Polyclonal antibody specifically detects Aprataxin in Human samples. It is validated for Western Blot
Specifications
| Aprataxin | |
| Polyclonal | |
| Western Blot | |
| AOA1EAOH, AOAMGC1072, aprataxin, ataxia 1, early onset with hypoalbuminemia, AXA1, EC 3.-, EOAHA, FHA-HIT, FLJ20157, Forkhead-associated domain histidine triad-like protein | |
| The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Aprataxin using the numbering given in SwissProt Entry Q7Z2E3 (GeneID 54840). | |
| 0.1 mL | |
| Cancer, DNA Repair, Tumor Suppressors | |
| 54840 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction