missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aprataxin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92693-0.02ml
This item is not returnable.
View return policy
Description
Aprataxin Polyclonal antibody specifically detects Aprataxin in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Aprataxin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunoprecipitation 1:20-1:50 | |
| AOA1EAOH, AOAMGC1072, aprataxin, ataxia 1, early onset with hypoalbuminemia, AXA1, EC 3.-, EOAHA, FHA-HIT, FLJ20157, Forkhead-associated domain histidine triad-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 93-342 of human APTX (NP_001182178.1). LYPYIVEFEEEAKNPGLETHRKRKRSGNSDSIERDAAQEAEAGTGLEPGSNSGQCSVPLKKGKDAPIKKESLGHWSQGLKISMQDPKMQVYKDEQVVVIKDKYPKARYHWLVLPWTSISSLKAVAREHLELLKHMHTVGEKVIVDFAGSSKLRFRLGYHAIPSMSHVHLHVISQDFDSPCLKNKKHWNSFNTEYFLESQAVIEMVQEAGRVTVRDGMPELLKLPLRCHECQQLLPSIPQLKEHLRKHWTQ | |
| 0.02 mL | |
| Cancer, DNA Repair, Tumor Suppressors | |
| 54840 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction