missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109293
This item is not returnable.
View return policy
Description
Positive Control: Human skin tissue lysates, A549, SH-SY-5Y, mouse liver tissue, MCF-7.
Promotes osteoblast cell differentiation and terminal mineralization. Plays a role in inducing the cell cycle arrest via inhibiting CCND1 expression in all-trans-retinoic acid (ATRA) signal pathway.
Specifications
| APR3 | |
| Polyclonal | |
| Unconjugated | |
| ATRAID | |
| 0610007C21Rik; 3-Apr; AI316792; all-trans retinoic acid induced differentiation factor; all-trans retinoic acid-induced differentiation factor; apoptosis related protein 3; apoptosis related protein APR-3; apoptosis-related protein 3; APR3; APR-3; APR--3; ATRAID; C11H2orf28; C17H2orf28; C2orf28; H1E6 protein; HSPC013; LOW QUALITY PROTEIN: all-trans retinoic acid-induced differentiation factor; p18; p18 protein; PRO240; RGD1311605; UNQ214/PRO240 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 381629, 51374 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6PGD0, Q6UW56 | |
| ATRAID | |
| Synthetic peptide within mouse APR3 aa 174-223. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction