Learn More
Applied Biosystems™ TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR)
TaqMan™ PreAmp Master Mix pre-amplifies small amounts of cDNA without introducing amplification bias to the sample.
£1008.00 - £3540.00
Specifications
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
Product Line TaqMan™
View More Specs
Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15344674
View Documents Promotion Details
Applied Biosystems™
4488593
200 Reactions
£3540.00
200 reactions
View alternative products
Save Up to 
»
10515405
View Documents Promotion Details
Applied Biosystems™
4391128
40 Reactions
£1008.00
Each
View alternative products
Save Up to 
»
Description
Description
  • Analyze mRNA from precious samples such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded tissues (FFPE)
  • Stretch as little as 1ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TaqMan™ Gene Expression assays
  • Analyze up to 100 gene expression targets with minimal hands-on time
Order Info
Guaranteed minimum shelf life is 60 days.
Specifications
Specifications
Primer-probe
qPCR
TaqMan™
Pre-amplification
High
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
PreAmp Master Mix
cDNA