missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Applied Biosystems™ SimpliAmp™ Strip Plate Tray Cover

Product Code. 17809152 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17809152 10 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17809152 Supplier Thermo Scientific™ Supplier No. A55971
  • ONLINE EXCLUSIVE PRICE
    £89.45
    £81.43 / Pack of 10
    Save £8.02
    8% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ease of sample setup and reduced handling steps enabled using the SimpliAmp Strip Plate Tray Cover.

Pipette and centrifuge with confidence with the Applied Biosystems™ SimpliAmp™ Strip Plate Tray Cover.

Used alongside the Thermo Scientific lysis tubes and pierceable seals to securely pipette from the Lysis Tubes and alongside the Thermo Scientific Assay Tubes, securely centrifuge with the Thermo Scientific™ SureTect™ PCR Assay Workflow enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.

Use the SimpliAmp Strip Plate Tray Cover to securely centrifuge SureTect PCR tubes and pipette from your SureTect lysis tubes in the SureTect workflow.

Designed to work in conjunction with the Thermo Scientific lysis and PCR tubes, SimpliAmp Strip Plate Tray and pierceable seals, the SimpliAmp Strip Plate Tray Cover enables you to streamline your SureTect workflow regardless of sample throughput.

Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.

We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Food
For Use With (Equipment) Thermo Scientific lysis and PCR tubes, SimpliAmp Strip Plate Tray and pierceable seals
Product Line SureTect
Quantity 10
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.