missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MuLV Reverse Transcriptase, Reduced
Description
Includes
100μL of MuLV Reverse Transcriptase (50U/μL)
- MuLV Reverse Transcriptase is a recombinant RNA-dependent DNA polymerase that uses single-stranded RNA as a template in the presence of a primer to synthesize a complementary DNA (cDNA) strand
- It can be used for the preparation of cDNA libraries, for first-strand synthesis in RT-PCR, or in dideoxy sequencing reactions in place of Klenow enzyme, for G+C–rich regions
- Optimal reaction temperature: 42° to 52°C
- Final product size: 5kb or less
PCR & Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Content And Storage | 100μL of MuLV Reverse Transcriptase (50U/μL). Store between -15deg. to -25deg.C. |
| Quantity | 5000 units |
| Sensitivity | High |
| For Use With (Application) | cDNA Synthesis, RT-PCR |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?