missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Multicolor Reaction Plates with Barcode

Ideal solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments

Brand:  Applied Biosystems™ 4483356

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15293005

  • £3310.00 / Pack of 500
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

  • Optimized for performance with Applied Biosystems instruments
  • Certified DNAse-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI™/SBS standard, well suited to high-throughput laboratories and robot handling
  • Great mechanical stability and flatness for complete workflow needs
  • Use of polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability
  • Ideal choice for use with automation/robotics and multi-instrument experiments (Like thermal cycler, real-time thermal cycler, genetic analyzer)
  • Thin-wall construction allows for snug fit to thermal blocks with excellent heat transfer from thermal block to sample
  • Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity

  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Specifications

Specifications

Includes Barcode
Store at room temperature.
MicroAmp
Microplate
Semi
0.3 mL
0.2 mL
Standard
Certified DNA/RNase Free
500 Plates
Optical, Assorted
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Polycarbonate, Polypropylene
96
500
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers