missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Blue Reaction Plates with Barcode

Product Code. 15233005 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
20 plates
Unit Size:
Pack of 20
Product Code. Quantity unitSize
15233005 20 plates Pack of 20
1 options
This item is not returnable. View return policy

Product Code. 15233005

Brand: Applied Biosystems™ 4483343

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for work that requires special handling and greater degree of durability for use with multi-instrument experiments

  • Optimized for performance with Applied Biosystems instruments
  • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI™/SBS standard, well suited to high-throughput laboratories and robot handling
  • Great mechanical stability and flatness for complete workflow needs
  • Polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and high durability
  • Ideal choice for use with automation/robotics and multi-instrument experiments (like thermal cycler, real-time thermal cycler, genetic analyzer)
  • Thin-wall construction allows for snug fit to thermal blocks with excellent heat transfer from thermal block to sample
  • Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity

  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage Store at room temperature.
Packaging 20 Plates
Product Line MicroAmp
Color Optical, Blue
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polycarbonate, Polypropylene
Volume (Metric) Well 0.3 mL
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 20 plates
Reaction Speed Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.