missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 8-Cap Strip, clear

Product Code. p-4926358 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1500 Strips (5 pk./300)
300 Strips
Unit Size:
Pack of 300
Pack of 8
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10209104

Brand: Applied Biosystems™ N8010535

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 8-cap strips available in clear or an assorted set of red, orange, blue, and green.

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce the possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
    • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
    • When ready, just peel away caps using tabs at end
    Colored for Easy Organization
    • Available in different colors to help track tubes during assay preparation
    • For real-time PCR applications,use MicroAmp Optical 8-Cap Strips (Mfr. No. 4323032) with flat-top profile

    PCR, PCR & Real-Time PCR

    TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance Certified DNA/Rnase Free
    Color Natural
    Quantity 300 Strips
    For Use With (Equipment) Thermal Cyclers
    Product Line MicroAmp™ 
    Material Polypropylene
    Content And Storage Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Natural (clear) color. Store at room temperature.
    Shipping Condition Room Temperature
    Closure Type Domed Cap
    No. of Wells 96
    Format 300 Strip Caps
    Product Type 8-Strip Cap
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.