missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Express-96 Standard Tip Combs
Shop All Thermo Scientific Products
Click to view available options
:
96 Standard Tip Combs
Description
Includes
100 combs
- Fit models 4400076 and 4400078
Animal Health, Animal Health Sample Preparation, Automated Nucleic Acid Purification, Automated Total RNA Isolation, Automated Viral DNA/RNA Purification, DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues, Total RNA from Bacterial Samples, Total RNA from Blood, Viral RNA/DNA Purification
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Filtered | Filtered |
| Tip Style | Standard |
| Quantity | 100 pieces |
| Content And Storage | Store at room temperature. |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | MagMAX Express-96 Standard Magnetic Particle Processor |
| Product Line | MagMAX |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction