missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MagMAX™ Express-96 Deep Well Plates
Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
100 to 1000 μL
Unit Size:
Each
Description
- Designed for use with MagMAX Express-96 Deep Well Magnetic Particle Processors
- Sample input volume for deep well configuration is 100 to 1000μL per well
Animal Health, Animal Health Sample Preparation, Automated Nucleic Acid Purification, Automated Total RNA Isolation, Automated Viral DNA/RNA Purification, DNA & RNA Purification & Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells & Tissues, Total RNA from Bacterial Samples, Total RNA from Blood, Viral RNA/DNA Purification
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Volume (Metric) | 100 to 1000 μL |
| For Use With (Equipment) | MagMAX Express-96 Deep Well Magnetic Particle Processor |
| Product Line | MagMAX |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction