missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit (with extra proteinase K and DNA binding beads)

Product Code. 15434783 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2,500 preps
500 Preps
Unit Size:
1 set
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434783 2,500 preps 25µg
15424783 500 Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434783 Supplier Applied Biosystems™ Supplier No. A25920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue.

Note: This product is identical to the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. No. A25597) except that it comes with additional proteinase K and DNA Binding Beads, which some customers find useful for their specific applications.

The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue. gDNA purified with the kit is ideal for qPCR applications on sensitive, scalable platforms such as the QuantStudio™ 12K Flex System with OpenArray™ Block (with AccuFill™ System). The magnetic bead–based purification format allows you to easily scale from processing 12 to 500 samples a day, making it the ideal choice for pharmacogenomics (PGx) studies.

Features of the MagMAX™ DNA Multi-Sample Ultra Kit include:
• Streamlined protocols for numerous non-invasive biological samples
• Optimized to provide DNA yields suitable for OpenArray™ and Ion AmpliSeq™ applications
• Packaged for high-throughput needs

One Kit for a Diverse Set of Sample Types
Why purchase multiple purification kits to isolate gDNA from different sample types? The MagMAX™ DNA Multi-Sample Ultra Kit eliminates the headache of ordering these extra kits because it has been optimized for use with a variety of biological samples. The kit is designed for high-throughput isolation of gDNA from 1 buccal swab, 50 to 350 μL of whole blood, up to 400 μL urine (without sample concentration), up to 150 μL saliva, 1-5 mL mouth rinse, blood spot cards, and mammalian tissue samples (up to 10 mg). The protocols and magnetic bead format make the kit fully amenable to automation.

*Please note that the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor with a 24 Deep-well Head is recommended for sample volumes greater than 50 μL.

Benefits of MagMAX™ Magnetic Bead–based Purification
Magnetic beads offer many benefits compared to other technologies for isolating gDNA. Beads bind gDNA more efficiently than glass fiber filters, resulting in higher and more consistent gDNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples.

Accessory Products
Proteinase K (Cat. No. A25561), DNA Binding Beads (Cat. No. A25562), Wash Solution 1 Concentrate (Cat. No. AM8504), and Wash Solution 2 Concentrate (Cat. No. AM8640) are components of the kit that may also be purchased separately.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 ng/μL to 3 ng/μL
Content And Storage • DNA Binding Beads, 10 x 8 mL (store at 2°C to 8°C, DO NOT FREEZE)
• DNA Elution Buffer 1, 5 x 25 mL (store at room temperature)
• DNA Elution Buffer 2, 5 x 25 mL (store at room temperature)
• Wash Solution 1 Concentrate, 5 x 80 mL* (store at room temperature)
• Wash Solution 2 Concentrate, 5 x 162 mL* (store at room temperature)
• Multi-Sample DNA Lysis Buffer, 5 x 100 mL (store at room temperature)
• Nuclease-Free Water, 5 x 100 mL (store at room temperature)
• PK Buffer, 5 x 96 mL (store at room temperature)
• Proteinase K, 10 x 4 mL (store at -5° to -30°C)
• Ribonuclease A (RNase A), 5 x 1.25 mL (store at -5° to -30°C)
*after addition of alcohol
Format Kit
Isolation Technology Magnetic Bead
Sample Type Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum), Tissue
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
No. of Reactions 2500 Preps
Product Line MagMAX
Product Type DNA Multi-Sample Ultra Kit
Quantity 2,500 preps
Target Genomic DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.