Learn More
Applied Biosystems™ MagMAX™ DNA Multi-Sample Ultra Kit (with extra proteinase K and DNA binding beads)
Shop All Applied Biosystems ProductsDescription
Note: This product is identical to the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. No. A25597) except that it comes with additional proteinase K and DNA Binding Beads, which some customers find useful for their specific applications.
The MagMAX™ DNA Multi-Sample Ultra Kit is optimized to isolate genomic DNA (gDNA) from a variety of samples, such as whole blood, buccal cells, saliva, urine, blood cards, mouth rinse, and tissue. gDNA purified with the kit is ideal for qPCR applications on sensitive, scalable platforms such as the QuantStudio™ 12K Flex System with OpenArray™ Block (with AccuFill™ System). The magnetic bead–based purification format allows you to easily scale from processing 12 to 500 samples a day, making it the ideal choice for pharmacogenomics (PGx) studies.
Features of the MagMAX™ DNA Multi-Sample Ultra Kit include:
• Streamlined protocols for numerous non-invasive biological samples
• Optimized to provide DNA yields suitable for OpenArray™ and Ion AmpliSeq™ applications
• Packaged for high-throughput needs
One Kit for a Diverse Set of Sample Types
Why purchase multiple purification kits to isolate gDNA from different sample types? The MagMAX™ DNA Multi-Sample Ultra Kit eliminates the headache of ordering these extra kits because it has been optimized for use with a variety of biological samples. The kit is designed for high-throughput isolation of gDNA from 1 buccal swab, 50 to 350 μL of whole blood, up to 400 μL urine (without sample concentration), up to 150 μL saliva, 1-5 mL mouth rinse, blood spot cards, and mammalian tissue samples (up to 10 mg). The protocols and magnetic bead format make the kit fully amenable to automation.
*Please note that the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor with a 24 Deep-well Head is recommended for sample volumes greater than 50 μL.
Benefits of MagMAX™ Magnetic Bead–based Purification
Magnetic beads offer many benefits compared to other technologies for isolating gDNA. Beads bind gDNA more efficiently than glass fiber filters, resulting in higher and more consistent gDNA yields. Additionally, because filters are not used, there is no risk of filter clogging due to cellular particulates in samples.
Accessory Products
Proteinase K (Cat. No. A25561), DNA Binding Beads (Cat. No. A25562), Wash Solution 1 Concentrate (Cat. No. AM8504), and Wash Solution 2 Concentrate (Cat. No. AM8640) are components of the kit that may also be purchased separately.
Specifications
Specifications
| Content And Storage | • DNA Binding Beads, 2 x 8 mL (store at 2°C to 8°C, DO NOT FREEZE) • DNA Elution Buffer 1, 1 x 25 mL (store at room temperature) • DNA Elution Buffer 2, 1 x 25 mL (store at room temperature) • Wash Solution 1 Concentrate, 1 x 80 mL* (store at room temperature) • Wash Solution 2 Concentrate, 1 x 162 mL* (store at room temperature) • Multi-Sample DNA Lysis Buffer, 1 x 100 mL (store at room temperature) • Nuclease-Free Water, 1 x 100 mL (store at room temperature) • PK Buffer, 1 x 96 mL (store at room temperature) • Proteinase K, 2 x 4 mL (store at -5° to -30°C) • Ribonuclease A (RNase A), 1 x 1.25 mL (store at -5° to -30°C) *after addition of alcohol |
| Format | Kit |
| Sample Type | Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum), Tissue |
| For Use With (Application) | Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR) |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| No. of Reactions | 500 Preps |
| Product Line | MagMAX |
| Product Type | DNA Multi-Sample Ultra Kit |
| Quantity | 500 Preps |
| Target | Genomic DNA |
Frequently Asked Questions (FAQs)
This master mix has a heat-labile UDG. It is completely and irreversibly inactivated after 10 minute incubation at 50°C.
We have optimized protocols for extracting DNA from buccal swabs, whole blood, blood cards (Whatman FTA), oral rinse, saliva, and urine samples. In addition, the kit can be used to process vaginal microbiota samples (e.g., ThinPrep, Surepath) for gDNA extraction from yeast, bacteria (Gram positive/negative), viruses, and protozoa with the addition of additional enzymatic steps.
On rare occasions, the sample may turn cloudy after adding the Multi-Sample DNA Lysis Buffer. The sample will clear up when isopropanol is added. This will not have any impact on the yield or quality of the DNA.
Heparin is not recommended as an anti-coagulant since it can cause inhibition of PCR reactions. Instead, we strongly recommend collecting blood samples in EDTA or sodium citrate.
We recommend transferring the lysate to a new MagMAX Express-96 Deep Well Plate (Cat. No. 4388476). This option eliminates contamination risks and saves time. To transfer lysates, set a multi-channel micropipetor to 300 µL and transfer one row at a time. Each well should contain 200-250 µL after transfer.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.