Learn More
Applied Biosystems™ GeneAmp™ Fast PCR Master Mix (2X), with or without protocol
Shop All Applied Biosystems ProductsDescription
Includes
The GeneAmp Fast PCR Master Mix (2X) contains two tubes (1.25mL each) sufficient for 125 20μL PCR reactions. The Fast PCR Master Mix can also be ordered with a protocol.
Applied Biosystems GeneAmp Fast PCR Master Mix reduces PCR reaction time to as little as 25 minutes when used with the Veriti 96-well thermal cycler. The sensitive, premixed hot-start chemistries of this master mix enable quick experiment setup and by generating greater than 90% 'A' overhang, the cloning process is further simplified.
GeneAmp Fast PCR Master Mix may be used to detect a single copy of a gene in 10 ng of human genomic DNA or as little as 10 copies of an HIV target.
Features
- Quick reaction setup with premixed reagents
- Gene detection with as little as 10 ng of human genomic DNA
- Reproducible, specific, and sensitive results
Notes
- The Applied Biosystems Fast PCR System, consisting of the premixed GeneAmp Fast PCR Master Mix, specially designed microplates and tubes, and the Veriti 96-well thermal cycler (0.1 mL or 0.2 mL), considerably reduces PCR reaction times.
- GeneAmp Fast PCR Master Mix is provided at a convienient 2X concentration with only the addition of DNA template and primers required.
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | • 2X GeneAmp Fast PCR Master Mix, 2 x 1.25 mL tubes • Protocol Store at 2°C to 8°C. |
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | Medium |
| Polymerase | AmpliTaq DNA Polymerase |
| Reaction Speed | Fast |
| Exonuclease Activity | 5' - 3' |
| Product Line | GeneAmp |
| Product Type | Fast PCR Master Mix |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.