Learn More
Applied Biosystems™ GeneAmp™ dNTP Blend (100 mM)
Shop All Applied Biosystems ProductsDescription
GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 µL each. Each kit contains 12 x 1 mL vials.
Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use
Specifications
Specifications
| Concentration | 100 mM |
| Content And Storage | GeneAmp dNTP Blend, 100 mM (12 x 1 mL) Store in a constant temperature freezer at –20°C. |
| Form | Liquid |
| Product Line | GeneAmp |
| Quantity | 12 x 1 mL |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Standard PCR |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.