Learn More
Description
GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 µL each. Each kit contains 12 x 1 mL vials.
Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use
Specifications
Specifications
| Concentration | 100 mM |
| Content And Storage | GeneAmp dNTP Blend, 100 mM (12 x 1 mL) Store in a constant temperature freezer at –20°C. |
| Form | Liquid |
| Product Line | GeneAmp |
| Quantity | 12 x 1 mL |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Standard PCR |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.