missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DNA Binding Beads for MagMAX™ DNA Multi-Sample Ultra Kit

Product Code. 15681822 Shop All Applied Biosystems Products
Click to view available options
Quantity:
8 mL
Unit Size:
8mL
This item is not returnable. View return policy

Product Code. 15681822

Brand: Applied Biosystems™ A25562

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These are the same DNA Binding Beads found in the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. No. A25597, A25598), available separately in case more DNA Binding Beads are required for a particular application.

These are the same DNA Binding Beads found in the MagMAX™ DNA Multi-Sample Ultra Kit (Cat. No. A25597, A25598), available separately in case more DNA Binding Beads are required for a particular application.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C). DO NOT FREEZE.
Format Kit
Sample Type Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum), Tissue
For Use With (Application) Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Product Type DNA Binding Bead
Quantity 8 mL
Target Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.