missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Centri-Spin™ 20 Columns

Product Code. 10300595
Change view
Click to view available options
Quantity:
50 columns
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10300595 50 columns Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10300595 Supplier Invitrogen™ Supplier No. 4367830

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Process samples faster than ultrafiltration columns

  • Centri-Spin™ columns provide excellent recovery of RNA and DNA fragments for a variety of purification applications. Use Centri-Spin columns to remove primers, probes, and dNTPs from DNA, RNA, PCR and end-labeled products
  • Rapid and efficient separations
  • Permits user to determine buffer composition
  • Convenient 20-50μL sample size
  • Dry gel format
  • Room Temperature Storage
  • Microcentrifuge format
  • Excellent Recovery
  • Centri-Spin columns provide excellent recovery of DNA fragments with sizes greater than 10, 20 or 40 base pairs (depending on kit), while removing >98% of salts, NTP's and other low-molecular-weight impurities. Proteins, peptides, and protein conjugates >5kD can also be efficiently separated/desalted from unwanted low-molecular-weight impurities
  • Designed for Performance
  • The column design is based on the description of gel filtration for the purification of DNA from nick translation reactions by Sambrook, et.al. Each Centri-Spin consists of a specially fritted microfuge tube, dry gel, a wash tube and a sample collection tube, all specifically designed for this purpose
  • Versatile
  • The column gel can be hydrated with either reagent-grade water or a suitable buffer, and spun in a microcentrifuge or swinging-bucket centrifuge to remove the intersititial fluid. After the sample is applied, spin the column again to remove the low-molecular-weight components, leaving a purified sample exchanged into the selected buffer

DNA & RNA Purification & Analysis, DNA Extraction, PCR Product Clean-Up

TRUSTED_SUSTAINABILITY

Specifications

Column Type Spin Column
Content And Storage Store at room temperature
Description Centri-Spin 20
Format Combs, Pre-filled Cartridge, Tube
Product Line Centri-Spin
Product Type Spin Column
Quantity 50 columns
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.