missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Centri-Spin™ 40 Columns
Description
- Centri-Spin™ columns provide excellent recovery of RNA and DNA fragments for a variety of purification applications. Use Centri-Spin columns to remove primers, probes, and dNTPs from DNA, RNA, PCR and end-labeled products
- Rapid and efficient separations
- Permits user to determine buffer composition
- Convenient 20-50μL sample size
- Dry gel format
- Room Temperature Storage
- Microcentrifuge format
- Excellent Recovery
- Centri-Spin columns provide excellent recovery of DNA fragments with sizes greater than 10, 20 or 40 base pairs (depending on kit), while removing >98% of salts, NTP's and other low-molecular-weight impurities. Proteins, peptides, and protein conjugates >5kD can also be efficiently separated/desalted from unwanted low-molecular-weight impurities
- Designed for Performance
- The column design is based on the description of gel filtration for the purification of DNA from nick translation reactions by Sambrook, et.al. Each Centri-Spin consists of a specially fritted microfuge tube, dry gel, a wash tube and a sample collection tube, all specifically designed for this purpose
- Versatile
- The column gel can be hydrated with either reagent-grade water or a suitable buffer, and spun in a microcentrifuge or swinging-bucket centrifuge to remove the intersititial fluid. After the sample is applied, spin the column again to remove the low-molecular-weight components, leaving a purified sample exchanged into the selected buffer
DNA & RNA Purification & Analysis, DNA Extraction, PCR Product Clean-Up
Specifications
Specifications
| Column Type | Spin Column |
| Content And Storage | Store at room temperature |
| Description | Centri-Spin 40 |
| Format | Combs, Pre-filled Cartridge, Tube |
| For Use With (Application) | Nucleic Acid Purification |
| Product Line | Centri-Spin |
| Product Type | Spin Column |
| Quantity | 50 columns |
| Shipping Condition | Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?