missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Centri-Sep™ Spin Columns
Click to view available options
Quantity:
100 Columns
32 Columns
Unit Size:
Pack of 100
Pack of 32
Description
- Centri-Sep™ Spin Columns use Centri-Sep™ gel that provides excellent recovery of DNA fragments larger than 16bp, while removing greater than 98% of salts, NTPs, and other low molecular weight impurities
- Features of Centri-Sep™ columns: Processes samples faster than ultrafiltration columns
- Achieves rapid and efficient separation
- Permits use of custom buffers
- Convenient 20–100μL sample size
- Dry gel format
- Room temperature storage
- Microcentrifuge format
- Designed for performance: The column design is based on the description of gel filtration for the purification of DNA from nick translation reactions by Sambrook, et.al. Each Centri-Sep; unit consists of a specially fritted microfuge tube, dry gel, a wash tube, and a sample collection tube, all specifically designed for this purpose
- Excellent recovery The Centri-Sep gel provides excellent recovery of DNA fragments with sizes greater than 16bp or a 25-mer, while removing >98% of salts, NTPs, and other unwanted low molecular weight impurities
- Versatile The column gel is hydrated with either reagent-grade water or a buffer of your choice, and spun in a microcentrifuge or swinging-bucket centrifuge to remove the intersititial fluid. Your sample is then applied to the column and spun again, leaving you not only with a sample purified through removal of low molecular weight components, but also exchanged into the buffer of your choice
DNA & RNA Purification & Analysis, DNA Extraction, Sequencing Reaction Purification
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store at room temperature |
| Format | Combs, Pre-filled Cartridge, Tube |
| Product Line | Centri-Sep |
| Quantity | 100 Columns |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction