missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase, LD (Low DNA) with Gold Buffer and MgCl2

Product Code. p-4919370 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1000 Units
250 Units
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10055114

Brand: Applied Biosystems™ 4338856

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Contains AmpliTaq Gold DNA Polymerase, LD (250 units), GeneAmp 10X PCR Gold Buffer, and MgCl2 Solution.

Applied Biosystems AmpliTaq Gold DNA Polymerase, LD (Low DNA) is a highly purified version of AmpliTaq Gold DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations.

Applied Biosystems AmpliTaq Gold DNA Polymerase, LD (Low DNA), is a highly purified version of AmpliTaq Gold DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations.

This highly purified enzyme preparation gives you more confidence in your results and makes advanced PCR techniques quick and easy. When combined with the supplied GeneAmp 10X PCR Gold Buffer, AmpliTaq Gold DNA Polymerase, LD, provides a flexible hot-start system that is suitable for all PCR applications that require the elimination of bacterial DNA contamination.

Features

  • Highly purified enzyme with lower levels of bacterial DNA
  • Minimized non-target, false-positive PCR products when amplifying bacterial sequences
  • Chemically modified hot-start technique eliminates biological contamination
  • 10X PCR Gold Buffer promotes fast enzyme activation and robust PCR amplification
  • Enhanced specificity with time-release PCR for low-copy target amplification
  • Improved yield and success rate for target amplification

Notes

  • AmpliTaq Gold DNA Polymerase, LD, is recommended for PCR applications that require low background levels of bacterial DNA.
  • See user's manual or package insert for limited label license and trademark information.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • AmpliTaq Gold DNA Polymerase, LD (5 U/μL), 1 x 50 μL
• 10X PCR Gold Buffer, 1 x 1.5 mL
• 25 mM MgCl2, 1 x 1.5 mL

Store at -15°C to -30°C.
Format Tube
GC-Rich PCR Performance Low
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Purity or Quality Grade LD (Low DNA)
Quantity 250 Units
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.