missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ 360 Master Mix
GREENER_CHOICE

Product Code. 15594324 Shop All Applied Biosystems Products
Change view
Click to view available options
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15594324 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15594324 Supplier Applied Biosystems™ Supplier No. 4398886

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contains everything required for successful PCR amplification in a convenient package—all components are premixed and premeasured

AmpliTaq Gold 360 Master Mix was designed for 360° coverage of a full range of targets. The master mix is supplied at 2X the recommended usage concentration for easy dilution when adding template and primers. Designed for convenience, the AmpliTaq Gold 360 PCR Master Mix scales to various reaction volumes for greater application and format flexibility. The main ingredient of the master mix is AmpliTaq Gold 360 DNA Polymerase but it also contains the world-class 360 GC Enhancer which can be added optionally for high GC–content templates. With the wide template coverage, optimization of PCR reaction conditions, is virtually eliminated with the use of AmpliTaq Gold 360 Master Mix.

  • Optimized for broadest range of targets from everyday to challenging
  • Unmatched sensitivity, specificity, and yield
  • Robust amplification of GC-rich sequences with market-leading 360 GC Enhancer
  • Achieves highest-quality sequencing data
  • Easy-to-use, premixed master mix
  • Less waste, sustainable packaging

Amplification of Bisulfite-Treated DNA, GC-Rich PCR, Hot Start PCR, Methylation Analysis, Methylation-Sensitive Mobility Shift Assay, PCR, PCR and Real-Time PCR, RNAi, Epigenetics and Non-Coding RNA Research, Routine PCR

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 50 mL 2X AmpliTaq Gold 360 Master Mix
• 1 x 10 mL 360 GC Enhancer

Store at -20°C until first use, then store at 4°C.
Format Tube
GC-Rich PCR Performance High
Polymerase AmpliTaq Gold 360 DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Green Features Sustainable packaging
Product Type PCR Master Mix
Quantity 50 mL
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
For Use With (Application) Cloning
Fidelity (vs. Taq) 1 X
Hot Start Built-In Hot Start
No. of Reactions 2000
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Size (Final Product) 5 kb or less
Starting Material DNA
Volume 50 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.