missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ 360 DNA Polymerase
GREENER_CHOICE

Product Code. 10269814 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 Units
1000 Units
1500 Units
25 x 1000 Units
250 Units
3000 Units
5000 Units
Unit Size:
25000 units
3000 units
5000 units
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10269814 1500 Units Each
10229234 100 Units Each
10695795 250 Units Each
10147004 1000 Units Each
10525405 25 x 1000 Units 25000 units
10137204 3000 Units 3000 units
10753647 5000 Units 5000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10269814 Supplier Applied Biosystems™ Supplier No. 4398892

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

AmpliTaq Gold 360 DNA Polymerase is supplied at 5U/μL with 10X AmpliTaq Gold 360 PCR Buffer, 25mM M2Cl2, and 360 GC Enhancer. Typically, 1.5mL of each of AmpliTaq Gold 360 Buffer, 25mM MgCl2, and 360 GC Enhancer is supplied with each 250U of enzyme. Storage Buffer Enzyme is supplied in 20mM Tris, pH 9.0, 100mM KCl, 0.1mM EDTA, 1mM DTT, 0.5% (w/v) Tween 20, 50% (v/v) glycerol. Source: Recombinant, modified form of theThermus aquaticusDNA polymerase gene expressed in E. coli. All solutions are prepared in deionized, filter-sterilized water.

The Applied Biosystems AmpliTaq Gold 360 DNA Polymerase, when used with improved AmpliTaq Gold 360 Buffer, and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts.

The Applied Biosystems AmpliTaq Gold 360 DNA Polymerase, when used with AmpliTaq Gold 360 Buffer and the optional 360 GC Enhancer, amplifies a vast range of DNA sequence contexts. This polymerase efficiently amplifies targets present at low copy number, even in the presence of high concentrations of complex DNA, making it especially suited for low-copy pathogen detection and amplification of targets from degraded DNA samples. AmpliTaq Gold 360 DNA Polymerase delivers 360° coverage for a full range of targets.

AmpliTaq Gold 360 DNA Polymerase is purified by a proprietary separation process to eliminate contaminating bacterial DNA sequences from the enzyme preparation. When used with the enhanced 360 Gold Buffer, this ultra-pure enzyme, in addition to its hot-start capabilities, reduces false positive results, amplifies low-level target sequences, and promotes the amplification of a variety of templates including those from bacterial and human genomes.

Features

  • Optimized for the broadest range of targets
  • High sensitivity, specificity, and yield
  • Robust amplification of GC-rich sequences with 360 GC Enhancer
  • High-quality sequencing data
  • Efficiently and reproducibly amplifies sequences up to 5 Kb

Optimized for easy and challenging targets

Challenging targets include AT-rich, GC-rich, primer-dimer forming amplicons, homopolymer repeats, and amplicons that pose sequencing challenges. Amplicons that previously required specialized enzymes and reaction conditions can now be reproducibly amplified with a single reagent under standardized conditions (see figure).

Optimized for hot-start PCR

AmpliTaq Gold 360 DNA Polymerase provides the same hot-start specificity as AmpliTaq Gold DNA Polymerase. A high-temperature incubation step is required to activate AmpliTaq Gold DNA Polymerase, which ensures that the active enzyme is generated only at temperatures in which the DNA is fully denatured and when the primers are not annealed.

When the polymerase is added to the reaction mixture at room temperature, primer extension does not occur because the enzyme is inactivated. Any low stringency mispriming events that may have occurred will not be enzymatically extended and will not be amplified. Hence, PCR setup can be performed at room temperature without concern for extension at misprimed sites. The amount of AmpliTaq Gold 360 DNA Polymerase increases in the reaction slowly with each cycle number, and specific product yield increases without buildup of nonspecific products, including primer dimers.

Notes

  • See figure below for a demonstration of robust PCR amplification of long human and plasmid DNA.
  • See user's manual or package insert for limited label license and trademark information.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • AmpliTaq Gold 360 DNA Polymerase (5 U/μL), 6 x 50 μL
• 10X AmpliTaq Gold 360 Buffer, 6 x 1.5 mL
• 25 mM MgCl2, 6 x 1.5 mL
• 360 GC Enhancer, 6 x 1.5 mL

Store at -15°C to -25°C.
Format Tube
GC-Rich PCR Performance High
Polymerase AmpliTaq Gold 360 DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Quantity 1500 Units
Shipping Condition Room Temp or Wet Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 1200 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.