Learn More
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I or Buffer II
Shop All Applied Biosystems ProductsDescription
Includes
Contains 1000 units of AmpliTaq DNA Polymerase, Buffer II, and MgCl2 Solution.
AmpliTaq DNA Polymerase is a 94-kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is the most thoroughly characterized enzyme available for the PCR process, and its recombinant nature and purification method provide unparalleled purity and reproducibility, vial-to-vial and lot-to-lot.
AmpliTaq DNA Polymerase is available with GeneAmp10X PCR Buffer I or with GeneAmp 10X PCR Buffer II and MgCl2 Solution.
Reliable and robust PCR
The thermal activity profile of AmpliTaq DNA Polymerase is good for PCR applications because its optimal activity is in the same range at which stringent annealing of primers occurs (55–75°C). The enzyme's half-life is ∼40 minutes at 95°C, providing thermostability that meets the requirements of most difficult PCR applications.
Features
- The most thoroughly characterized enzyme available for the PCR process
- Excellent thermal activity profile makes it good for PCR applications
- QC-tested to guarantee reproducible results
Notes
- For superior PCR performance, we recommend DreamTaq DNA Polymerase.
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Concentration | 5 U/μL |
| Content And Storage | • AmpliTaq DNA Polymerase (5 U/μL), 25 x 200 μL • GeneAmp 10X PCR Buffer II, 100 x 1.5 mL • 25 mM MgCl2, 100 x 1.5 mL Store at -20°C. |
| Format | Stand-alone enzyme |
| Polymerase | AmpliTaq DNA Polymerase |
| Reaction Speed | Standard |
| Exonuclease Activity | 5' - 3' |
| Product Type | DNA Polymerase |
| Quantity | 25,000 units |
| Shipping Condition | Dry Ice |
| For Use With (Application) | Standard PCR |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.