missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-76565-25ul
This item is not returnable.
View return policy
Description
APPL Polyclonal specifically detects APPL in Human samples. It is validated for Western Blot.
Specifications
| APPL | |
| Polyclonal | |
| Western Blot 0.4 μg/mL | |
| Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26060.0 | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| APPL1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KIALLEPLLGYMQAQISFFKMGSENLNEQLEEFLANIGTSVQNVRREMDSDIETMQQTIEDLEVASDPLYVPDPDPTKFPVNR | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction