missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPBP1 Antibody (OTI1E10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72930F
This item is not returnable.
View return policy
Description
APPBP1 Monoclonal specifically detects APPBP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| NEDD8 Activating Enzyme (APPBP1/UBA3) | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NAE1 | |
| Recombinant protein fragment corresponding to amino acids 1-274 of human NEDD8 Activating Enzyme (APPBP1/UBA3) (NP_001018170) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI1E10 | |
| Immunohistochemistry | |
| A-116A10.1, amyloid beta precursor protein binding protein 1, amyloid beta precursor protein binding protein 1, 59kDa, Amyloid beta precursor protein-binding protein 1, 59 kDa, amyloid beta precursor protein-binding protein 1, 59kD, Amyloid protein-binding protein 1, APPBP1APP-BP1, NEDD8 activating enzyme E1 subunit 1, NEDD8-activating enzyme E1 regulatory subunit, NEDD8-activating enzyme E1 subunit, protooncogene protein 1, Proto-oncogene protein 1, ula-1 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 8883.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction