missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APPBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38082-100ul
This item is not returnable.
View return policy
Description
APPBP1 Polyclonal antibody specifically detects APPBP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| APPBP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| A-116A10.1, amyloid beta precursor protein binding protein 1, 59kDa, Amyloid beta precursor protein-binding protein 1, 59 kDa, amyloid beta precursor protein-binding protein 1, 59kD, APP-BP1, HPP1, NAE1, NEDD8 activating enzyme E1 subunit 1, NEDD8-activating enzyme E1 regulatory subunit, NEDD8-activating enzyme E1 subunit, protooncogene protein 1, Proto-oncogene protein 1, ula-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human APPBP1 (NP_003896.1).,, Sequence:, MAQLGKLLKEQKYDRQLRLWGDHGQEALESAHVCLINATATGTEILKNLVLPGIGSFTIIDGNQVSGEDAGNNFFLQRSSIGKNRAEAAMEFLQELNSDVSGSFVEESPENLLDNDPSFFCRFTVVVATQLPESTSLRLADVLWNSQIPLLICRTYGLVGYMRIIIKEHPVIESHPDNALEDLRLDKPFPELREHFQSYDLDHMEKKDHSHTPWIVIIAK | |
| 100 μL | |
| Apoptosis | |
| 8883 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction