missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APP Rabbit anti-Human, Mouse, Rat, Clone: 3A8S2, Novus Biologicals™
Rabbit Monoclonal Antibody
£173.00 - £415.00
Specifications
| Antigen | APP |
|---|---|
| Clone | 3A8S2 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18328154
|
Novus Biologicals
NBP3-15560-20UL |
20 μg |
£173.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18030126
|
Novus Biologicals
NBP3-15560-100UL |
100 μg |
£415.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APP Monoclonal antibody specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDownSpecifications
| APP | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP | |
| A synthetic peptide corresponding to a sequence within amino acids 671-770 of human APP (P05067). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3A8S2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 351 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title