missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
APP Monoclonal antibody specifically detects APP in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | APP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI7G9 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human APP (NP_000475) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?