missing translation for 'onlineSavingsMsg'
Learn More

Apovincaminic Acid Ethyl Ester N-Oxide, TRC

Product Code. 30330510
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330510 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330510 Supplier Toronto Research Chemicals Supplier No. TRCA729255250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinpocetine N-Oxide
CAS 109741-24-4
Molecular Formula C22 H26 N2 O3
Quantity 250 mg
Synonym Eburnamenine-14-carboxylic acid, ethyl ester, 4-oxide, (3α,16α)- (9CI), 1H-Indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine, eburnamenine-14-carboxylic acid deriv., Apovincaminic acid ethyl ester N-oxide, Ethyl apovincaminate N-oxide
InChI Formula InChI=1S/C22H26N2O3/c1-3-22-11-7-12-24(26)13-10-16-15-8-5-6-9-17(15)23(19(16)20(22)24)18(14-22)21(25)27-4-2/h5-6,8-9,14,20H,3-4,7,10-13H2,1-2H3/t20-,22+,24?/m1/s1
SMILES CCOC(=O)C1=C[C@]2(CC)CCC[N+]3([O-])CCc4c([C@H]23)n1c5ccccc45
IUPAC Name ethyl (15S,19S)-15-ethyl-11-oxido-1-aza-11-azoniapentacyclo[9.6.2.02,7.08,18.015,19]nonadeca-2,4,6,8(18),16-pentaene-17-carboxylate
Molecular Weight (g/mol) 366.4534
Formula Weight 366.1943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.