missing translation for 'onlineSavingsMsg'
Learn More

Apoptozole, MedChemExpress

Product Code. 16492427
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492427 50 mg 50mg
16478897 5 mg 5mg
16426318 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492427 Supplier Medchem Express Supplier No. HY15098
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Apoptozole (Apoptosis Activator VII) is an inhibitor of the ATPase domain of Hsc70 and Hsp70, with Kds of 0.21 and 0.14 μM, respectively, and can induce apoptosis.

Apoptozole (Apoptosis Activator VII) is an inhibitor of the ATPase domain of Hsc70 and Hsp70, with Kds of 0.21 and 0.14 μM, respectively, and can induce apoptosis.

Specifications

Chemical Name or Material Apoptozole
CAS 1054543-47-3
Color White
Purity Grade Notes Research
Molecular Formula C33H25F6N3O3
Quantity 50 mg
Synonym Apoptosis Activator VII
Solubility Information DMSO : ≥ 100 mg/mL (159.86 mM)
SMILES NC(C(C=C1)=CC=C1CN2C(C3=CC=C(OC)C=C3)=C(C4=CC=C(OC)C=C4)N=C2C5=CC(C(F)(F)F)=CC(C(F)(F)F)=C5)=O
Molecular Weight (g/mol) 625.56
Formula Weight 625.56
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.