missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apoptosis-Enhancing Nuclease Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522208
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HepG2, Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Apoptosis-Enhancing Nuclease (AEN) is a nuclear exonuclease that serves as a critical player in p53-mediated apoptosis. Induced by p53 in response to various DNA damage signals, AEN is involved in the degradation of cellular DNA during apoptosis. The nuclease activity of AEN contributes to the controlled dismantling of cellular components, ensuring that the apoptotic process leads to the efficient and timely removal of damaged or unwanted cells. Its expression regulation via phosphorylation highlights the intricacy of apoptotic signaling pathways and underscores the importance of AEN in maintaining genomic stability and preventing tumorigenesis.
Specifications
| Apoptosis-Enhancing Nuclease | |
| Polyclonal | |
| Unconjugated | |
| AEN | |
| 2700083B06Rik; AEN; aen {ECO:0000250; apoptosis enhancing nuclease; apoptosis-enhancing nuclease; apoptosis-enhancing nuclease {ECO:0000250; interferon stimulated exonuclease gene 20kDa-like 1; interferon stimulated exonuclease gene 20-like 1; interferon-stimulated 20 kDa exonuclease-like 1; interferon-stimulated 20 kDa exonuclease-like 1 {ECO:0000312; ISG20L1; pp12744; RGD:1305051}; RGD1305051; SBBI58; UniProtKB:Q8WTP8} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.9 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WTP8 | |
| AEN | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Apoptosis-enhancing nuclease. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction