missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Apoptosis Activator 2
GREENER_CHOICE

Product Code. 18044832
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18044832 50 mg 50mg
18025102 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18044832 Supplier Tocris Bioscience™ Supplier No. 2098/50
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Promotes apoptosome formation and activates caspase-9/caspase-3 pathway. Selectively induces tumor cell apoptosis

Apoptosis activator; promotes the cytochrome c-dependent oligomerization of Apaf-1 into the mature apoptosome. Increases procaspase-9 processing and subsequent caspase-3 activation. Induces apoptosis in tumor cells (IC50 = 4 - 9 μM for leukemia cells) with weak or no effect on normal cell lines or those defective/deficient in Apaf-1, caspase-9 or caspase-3 activity (IC50 > 40 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79183-19-0
Molecular Formula C15H9Cl2NO2

Specifications

Quantity 50 mg
Formula Weight 306.14
Purity >99%
Molecular Weight (g/mol) 306.14
Chemical Name or Material Apoptosis Activator 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.