missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APOOL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145132
This item is not returnable.
View return policy
Description
Immunogen sequence: QLPIYTAPPL QSKYVEEQPG HLQMGFASIR TATGCYIGWC KGVYVFVKNG IMDTVQFGKD AYVYLKNPPR DFLPKMGVIT VSGLAGLVSA RKGSKFKK Antigen sequence identity: Mouse 85%; Rat 84%.
This gene encodes a protein which contains an apolipoprotein O superfamily domain. This domain is found on proteins in circulating lipoprotein complexes. [provided by RefSeq, Sep 2011].
Specifications
| APOOL | |
| Polyclonal | |
| Unconjugated | |
| APOOL | |
| 6720473G16Rik; 9430083G14Rik; AAIR8193; apolipoprotein O like; apolipoprotein O-like; Apool; CXorf33; E130106L15Rik; FAM121A; family with sequence similarity 121A; MIC27; MICOS complex subunit MIC27; Protein FAM121A; RGD1549725; UNQ8193; UNQ8193/PRO23204 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 139322 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXV4 | |
| APOOL | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 40-137. Recombinant protein control fragment (Product #RP-110241). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction