missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12817-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.Specifications
| APOM | |
| Polyclonal | |
| Unconjugated | |
| APOM | |
| Apo M, apolipoprotein M, APOM, G3a, HSPC336, NG20, Protein G3a | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55937, 55938, 55939 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95445, P14630, Q9Z1R3 | |
| APOM | |
| APOM Fusion Protein Ag3137 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction