missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Apolipoprotein L4 Recombinant Protein Antigen

Product Code. 18158893 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18158893 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18158893 Supplier Novus Biologicals™ Supplier No. NBP214302PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human APOL4. The Apolipoprotein L4 Recombinant Protein Antigen is derived from E. coli. The Apolipoprotein L4 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-14302. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 80832
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol APOL4
Label Type Unlabeled
Molecular Weight (g/mol) 27kDa
Product Type Apolipoprotein L4
Quantity 0.1 mL
Regulatory Status RUO
Research Category metabolism
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-14302. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen TSTDQLEALRDILRDITPNVLSFALDFDEATKMIANDVHTLRRSKATVGRPLIAWRYVPINVVETLRTRGAPTRIVRKVARNL
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.