missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L3 Polyclonal Antibody

Apolipoprotein L3 Polyclonal Antibody
Brand: Thermo Scientific PA5103610
This item is not returnable.
View return policy
Description
Apolipoprotein L3 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| Apolipoprotein L3 | |
| Polyclonal | |
| Unconjugated | |
| APOL3 | |
| APOL3; APOLIII; ApoL-III; apolipoprotein L, 3; Apolipoprotein L3; apolipoprotein L-III; CG12_1; CG121; CG12-1; OTTHUMP00000028913; TNF-inducible protein CG12-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80833 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95236 | |
| APOL3 | |
| A synthesized peptide derived from human APOL3(Accession O95236), corresponding to amino acid residues D49-L99. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction