Learn More
Invitrogen™ Apolipoprotein L3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589476
Description
Immunogen sequence: MDSEKKRFTE EATKYFRERV SPVHLQILLT NNEAWKRFVT AAELPRDEAD ALYEALKKLR TYAAIEDEYV QQKDEQFREW FLKEFPQVKR KIQESIEKLR ALANGIEEVH RGCTISNVVS SSTGAASGIM SLAGLVLAPF TAGTSLALTA AGVGLGAASA VTGITTSIVE HSYTSSAEAE ASRLTATSID RLKVFKEVMR DITPNLLSLL NNYYEATQTI GSEIRAIRQA RARARLPVTT WRISAGSGGQ AERTIAGTTR AVSRGARILS ATTSGIFLAL DVVNLVYESK HLHEGAKSAS AEELRRQAQE LEENLMELTQ IYQRLNPCHT H; Positive Samples: DU145, A-549, BxPC-3, Mouse kidney; Cellular Location: Cytoplasm.
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein L gene family encodes six highly homologous proteins designated apoL1-6, which are associated with large high density type lipoproteins (HDL). The human apoL family maps to chromosome 22q12.1-13.1 within a 127,000 bp region. apoL2 and apoL3 may allow the binding of lipids to organelles or may be involved in the movement of lipids in the cytoplasm. apoL2 localizes to the cytoplasm and is widely expressed, with the highest levels observed in the lung, thymus, pancreas, placenta, adult brain and prostate. The apoL2 protein is also detected in the spleen, liver, kidney, colon, small intestine, uterus, spinal cord, adrenal gland, salivary gland, trachea, mammary gland, skeletal muscle, testis and fetal brain and liver. apoL3 also localizes to the cytoplasm and is widely expressed, with highest levels detected in the prostate, lung and placenta. It is also detected in kidney, bone marrow, spleen, thymus, spinal cord, adrenal gland, salivary gland, trachea and mammary gland. Lower levels of apoL3 are observed in the brain, heart, fetal liver, pancreas and testis.
Specifications
| Apolipoprotein L3 | |
| Polyclonal | |
| Unconjugated | |
| APOL3 | |
| APOL3; APOLIII; ApoL-III; apolipoprotein L, 3; Apolipoprotein L3; apolipoprotein L-III; CG12_1; CG121; CG12-1; OTTHUMP00000028913; TNF-inducible protein CG12-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80833 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.75 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O95236 | |
| APOL3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human APOL3 (NP_6636141). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.