missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98660AF700
This item is not returnable.
View return policy
Description
Apolipoprotein L2 Polyclonal specifically detects Apolipoprotein L2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Apolipoprotein L2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 23780 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| APOL3, APOL-II, apolipoprotein L, 2, apolipoprotein L2, Apolipoprotein L-II | |
| Produced in rabbits immunized with E. coli-derived Human Apolipoprotein L2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction