missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L1 Rabbit anti-Human, Mouse, Clone: 1A8N2, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | Apolipoprotein L1 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1A8N2 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 303-398 of human Apolipoprotein L1 (O14791). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?