missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Apolipoprotein L1 Monoclonal Antibody (CL0170)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531325

Product Code. 16324774

  • £480.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SNFLSLAGNT YQLTRGIGKD IRALRRARAN LQSVPHASAS RPRVTEPISA ESGEQVERVN EPSILEMSRG VKLTDVAPVS FFLVLDVVYL VYESKHLHEG AKSETAEELK KVAQELEEKL NILNN Highest antigen sequence indentity to the following orthologs: Mouse - 43%, Rat - 39%.

APOL1 is a secreted high density lipoprotein which binds to apolipoprotein A-I. Apolipoprotein A-I is a relatively abundant plasma protein and is the major apoprotein of HDL. It is involved in the formation of most cholesteryl esters in plasma and also promotes efflux of cholesterol from cells. This apolipoprotein L family member may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Apolipoprotein L1
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14791
APOL1
Recombinant protein fragment of Human APOL1 (Product #RP-100508)
100 μL
Primary
Human
Antibody
IgG1
Western Blot
CL0170
Unconjugated
APOL1
APOL; APO-L; APOL- I; APOL1; APOL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L1; Apolipoprotein L-I; FSGS4; OTTHUMP00000028705
Mouse
Protein A
RUO
8542
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.