missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein H/ApoH Antibody (OTI4A11), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70186AF350
This item is not returnable.
View return policy
Description
Apolipoprotein H/ApoH Monoclonal specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Apolipoprotein H/ApoH | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| APOH | |
| Recombinant protein fragment corresponding to amino acids 120-345 of human Apolipoprotein H/ApoH (NP_000033) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4A11 | |
| Western Blot, Immunohistochemistry | |
| Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG | |
| Mouse | |
| Affinity Purified | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 350.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction