missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein H/ApoH Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30735230 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735230 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735230 Supplier Novus Biologicals Supplier No. AF5087MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Apolipoprotein H/ApoH Polyclonal antibody specifically detects Apolipoprotein H/ApoH in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein H/ApoH
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias Activated protein C-binding protein, Anticardiolipin cofactor, APC inhibitor, Apo-H, Apolipoprotein H, apolipoprotein H (beta-2-glycoprotein I), B2G1, B2GP1, B2GPI, Beta(2)GPI, beta-2-glycoprotein 1, Beta-2-glycoprotein I, BG
Host Species Goat
Immunogen E. coli-derived recombinant human Apolipoprotein H/ApoH, Gly20-Cys345, Accession # P02749
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 350
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.