Learn More
Invitrogen™ Apolipoprotein F Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101430
Product Code. 16677904
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total APOF.
The product of this gene is one of the minor apolipoproteins found in plasma. This protein forms complexes with lipoproteins and may be involved in transport and/or esterification of cholesterol.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Apolipoprotein F
Polyclonal
Unconjugated
APOF
AI255964; APOF; Apo-F; Apolipoprotein F; APON; Ba1-666; DKFZp781G18150; Leukemia virus-inactivating factor; lipid transfer inhibitor protein; Liver regeneration-related protein LRRG151; LTIP; LVIF; MGC22520; ovarian and testicular apolipoprotein N; Unknown (protein for MGC:137251)
Rabbit
Affinity Chromatography
RUO
319
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q13790
APOF
A synthesized peptide derived from human APOF(Accession Q13790), corresponding to amino acid residues L293-I326.