missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E3/ApoE3 Antibody (960319), Alexa Fluor 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB41442AF532
This item is not returnable.
View return policy
Description
Apolipoprotein E3/ApoE3 Monoclonal specifically detects Apolipoprotein E3/ApoE3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
Specifications
| Apolipoprotein E3/ApoE3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| APOE | |
| ApoE3 variant peptide found within amino acids 100-200. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 960319 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry | |
| AD2, Apo-E, ApoE4, apolipoprotein E, LDLCQ5, LPG | |
| Mouse | |
| Protein A or G purified | |
| Lipid and Metabolism | |
| 348.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction