missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Apolipoprotein E R2/ApoE R2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92026-0.1ml
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Apolipoprotein E R2/ApoE R2 Polyclonal antibody specifically detects Apolipoprotein E R2/ApoE R2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Tekniske data
| Apolipoprotein E R2/ApoE R2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1 | |
| A synthetic peptide corresponding to a sequence within amino acids 900 to the C-terminus of human Apolipoprotein E R2/ApoE R2 (NP_004622.2). PLWAEPCLGETREPEDPAPALKELFVLPGEPRSQLHQLPKNPLSELPVVKSKRVALSLEDDGLP | |
| 0.1 mL | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 7804 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion