missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Apolipoprotein E Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHAPOE

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15454903

  • £528.00 / 1 set
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The Human Apolipoprotein E (Hu ApoE) ELISA quantitates Hu Apoe in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoE. Principle of the method The Human ApoE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13.2.
Specifications

Specifications

P02649
1.5 ng/mL
Apolipoprotein E (AD2) ELISA Kit, 96 Assays, 1 Kit, Human Species, HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of Apolipoprotein E (AD2) in Samples Using a Microplate Reader
Solid-phase sandwich ELISA
Colorimetric
GPCR (G-Protein Coupled Receptors)
This ELISA pair antibody detects human ApoE (isoforms ApoE2, ApoE3 and ApoE4).
Human
Colorimetric Microplate Reader
AD2,APO-E,ApoE4,LDLCQ5,LPG
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Anti-human Apo E precoated 96-well strip plate, lyophilized recombinant human Apo E standard, 20X wash buffer, biotinylated antibody reagent, streptavidin-HRP reagent, TMB substrate, stop solution, contains 0.2M sulfuric acid, assay diluent (5X concentrat
96 Tests
Cardiovascular Research
2°C to 8°C
Human
4 hr. 45 min.
1.64-400 ng/mL
1.5ng/mL to 400ng/mL
Apolipoprotein E (AD2), human ApoE (isoforms ApoE2, ApoE3 and ApoE4)
Biotin
ELISA Kit
Plasma, Serum, Supernatant
96 Assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
348
Human
<10%
HRP
RUO
Plasma, 1 μL; Serum, 1 μL; Supernatant, 100 μL
AD2, APO-E, LDLCQ5, LPG
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers